o-Anisaldehyde ≥98.0%
Supplier: TCI America
A0479-25G
A0479-500G
A0479-100G
TCA0479-025GEA
37.15
CAD
TCA0479-025G
TCA0479-500G
TCA0479-100G
o-Anisaldehyde ≥98.0%
o-Anisaldehyde
CAS Number: 135-02-4
MDL Number: MFCD00003308
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 243
Flash Point (°C): 118
Freezing point (°C): 36
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: 2-Methoxybenzaldehyde, Salicylaldehyde Methyl Ether
Formula:
C₈H₈O₂ Boiling Pt: 243 °C Melting Pt: 34…37 °C Density: 1.127 g/cm³ (25 °C) Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003308 CAS Number: 135-02-4 EINECS: 205-171-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...