m-Anisaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 3-Methoxybenzaldehyde

A0478-500ML A0478-25ML A0478-100ML
TCA0478-500MLEA 422.7 CAD
TCA0478-500ML TCA0478-025ML TCA0478-100ML
m-Anisaldehyde ≥98.0%
m-Anisaldehyde

CAS Number: 591-31-1
MDL Number: MFCD00003361
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 121
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: 3-Methoxybenzaldehyde

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 230 °C
Density: 1.12 (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1
EINECS: 209-712-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=193bb14af59f5cc8e9ad0519382265417c12b167&vid=05b16461bdea8f5103bb8592abd4651806e69d39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR