2-Amino-5-mercapto-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-Amino-1,3,4-thiadiazole-2-thiol

A0425-25G A0425-500G
TCA0425-025GEA 36.34 CAD
TCA0425-025G TCA0425-500G
2-Amino-5-mercapto-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-mercapto-1,3,4-thiadiazole

CAS Number: 2349-67-9
MDL Number: MFCD00003108
Molecular Formula: C2H3N3S2
Molecular Weight: 133.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 234


Other Product Names / Synonyms: 5-Amino-1,3,4-thiadiazole-2-thiol

Formula: C₂H₃N₃S₂
MW: 133.2 g/mol
Melting Pt: 234 °C
MDL Number: MFCD00003108
CAS Number: 2349-67-9
EINECS: 219-078-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef571913ad3be1cd236d03b8339361e7c7f8e742&vid=ef571913ad3be1cd236d03b8339361e7c7f8e742&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR