3-Aminoquinoline ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Quinolin-3-amine, Quinolin-3-ylamine
A0418-5G
A0418-25G
TCA0418-005GEA
101.12
CAD
TCA0418-005G
TCA0418-025G
3-Aminoquinoline ≥99.0%
3-Aminoquinoline
CAS Number: 580-17-6
MDL Number: MFCD00006772
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 137
Melting point (°C): 93
Formula:
C₉H₈N₂ Boiling Pt: 137 °C Melting Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006772 CAS Number: 580-17-6 EINECS: 209-455-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...