2-(4-Aminophenyl)ethanol ≥98.0%

Supplier: TCI America

Synonyms: 4-Aminophenethyl alcohol

A0394-25G A0394-100G
TCA0394-025GEA 141.43 CAD
TCA0394-025G 76218-702
2-(4-Aminophenyl)ethanol ≥98.0%
2-(4-Aminophenyl)ethanol

CAS Number: 104-10-9
MDL Number: MFCD00007922
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 255
Melting point (°C): 108


Other Product Names / Synonyms: 4-Aminophenethyl Alcohol, 2-(4-Aminophenyl)ethyl Alcohol

Formula: C₈H₁₁NO
Boiling Pt: 255 °C
Melting Pt: 108 °C
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92d5a4180fdb311289185de1de1e8f4b47cd04e9&vid=92d5a4180fdb311289185de1de1e8f4b47cd04e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR