4-Nitro-3-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Amino-2-nitro-α,α,α-trifluorotoluene, 4-Nitro-α,α,α-trifluoro-m-toluidine, 5-Amino-2-nitrobenzotrifluoride

A0377-25G A0377-5G
TCA0377-25GEA 96.38 CAD
TCA0377-25G TCA0377-005G
4-Nitro-3-(trifluoromethyl)aniline ≥98.0%
4-Nitro-3-(trifluoromethyl)aniline

CAS Number: 393-11-3
MDL Number: MFCD00014717
Molecular Formula: C7H5F3N2O2
Molecular Weight: 206.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Nitro-3-trifluoromethylaniline

Formula: C₇H₅F₃N₂O₂
MW: 206.12 g/mol
Melting Pt: 129
Density: 1.752 g/cm³ (–123.15 °C)
MDL Number: MFCD00014717
CAS Number: 393-11-3
EINECS: 206-884-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70144ed2aa1fd76a0e7a68b2590bfa24c9c41526&vid=70144ed2aa1fd76a0e7a68b2590bfa24c9c41526&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR