5-Amino-2-naphthalenesulfonic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A0346-500G A0346-25G
TCA0346-500GEA 244.58 CAD
TCA0346-500G TCA0346-025G
5-Amino-2-naphthalenesulfonic acid ≥97.0% (by titrimetric analysis)
5-Amino-2-naphthalenesulphonic acid

CAS Number: 119-79-9
MDL Number: MFCD00004030
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1,6-Cleve's Acid, 1-Naphthylamine-6-sulfonic Acid

Formula: C₁₀H₉NO₃S
MDL Number: MFCD00004030
CAS Number: 119-79-9
EINECS: 204-351-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31cef5894840426a82e876045fe87cbdbad38c1e&vid=31cef5894840426a82e876045fe87cbdbad38c1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR