2-Amino-p-cresol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline

A0330-500G A0330-25G A0330-100G
TCA0330-500GEA 801.69 CAD
TCA0330-500G TCA0330-025G TCA0330-100G
2-Amino-p-cresol ≥98.0% (by GC, titration analysis)
2-Amino-p-cresol

CAS Number: 95-84-1
MDL Number: MFCD00007699
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 135


Other Product Names / Synonyms: 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 135 °C
MDL Number: MFCD00007699
CAS Number: 95-84-1
EINECS: 202-457-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79124c1e313f254c348db95b8fe5ee8101e7037f&vid=79124c1e313f254c348db95b8fe5ee8101e7037f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR