Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,0

Supplier: TCI America
Warning

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

A0321-25G A0321-500G
TCA0321-025GEA 35.39 CAD
TCA0321-025G TCA0321-500G
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,0
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

CAS Number: 77-86-1
MDL Number: MFCD00004679
Molecular Formula: C4H11NO3
Molecular Weight: 121.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 170


Other Product Names / Synonyms: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trimethylolaminomethane, Tris-Base

Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357
Melting Pt: 170 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=111a48de13c37afd5f6305de8c40279b3a234cdd&vid=111a48de13c37afd5f6305de8c40279b3a234cdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR