4-Aminohippuric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0313-10G A0313-25G
TCA0313-010GEA 316 CAD
TCA0313-010G TCA0313-025G
4-Aminohippuric acid ≥99.0% (by titrimetric analysis)
4-Aminohippuric acid

CAS Number: 61-78-9
MDL Number: MFCD00007890
Molecular Formula: C9H10N2O3
Molecular Weight: 194.19
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 197


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(4-Aminobenzoyl)glycine

Formula: H₂NC₆H₄C(O)NHCH₂COOH
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 197 °C
Density: 1.402 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007890
CAS Number: 61-78-9
EINECS: 200-518-9
Merck Index: 13,00441
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65b7d505f14af451157dd32c82cfd20bb0307488&vid=65b7d505f14af451157dd32c82cfd20bb0307488&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR