Tryptamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A0300-1G
A0300-25G
TCA0300-001GEA
28.95
CAD
TCA0300-001G
TCA0300-025G
Tryptamine hydrochloride ≥98.0% (by titrimetric analysis)
Tryptamine hydrochloride
CAS Number: 343-94-2
MDL Number: MFCD00012682
Molecular Formula: C10H12N2
Molecular Weight: 196.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 248
Other Product Names / Synonyms: 3-(2-Aminoethyl)indole Hydrochloride
Formula:
C₁₀H₁₃ClN₂ Melting Pt: 248 °C |
MDL Number:
MFCD00012682 CAS Number: 343-94-2 EINECS: 206-446-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...