Ethanolamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0298-25G A0298-500G A0298-100G
TCA0298-025GEA 53.06 CAD
TCA0298-025G TCA0298-500G TCA0298-100G
Ethanolamine hydrochloride ≥98.0% (by titrimetric analysis)
Ethanolamine hydrochloride

CAS Number: 2002-24-6
MDL Number: MFCD00012892
Molecular Formula: C2H7NO
Molecular Weight: 97.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: Ethanolamine Hydrochloride, 2-Hydroxyethylamine Hydrochloride, Monoethanolamine Hydrochloride

Formula: C₂H₈ClNO
Melting Pt: 84 °C
MDL Number: MFCD00012892
CAS Number: 2002-24-6
EINECS: 217-900-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7813253f0c90bd4375e654d764e68dec7872d2f2&vid=7813253f0c90bd4375e654d764e68dec7872d2f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR