2-Aminobenzothiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

A0277-25G A0277-500G A0277-100G
TCA0277-025GEA 113.85 CAD
TCA0277-025G TCA0277-500G TCA0277-100G
2-Aminobenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Aminobenzothiazole

CAS Number: 136-95-8
MDL Number: MFCD00005785
Molecular Formula: C7H6N2S
Molecular Weight: 150.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Boiling Pt: 190…195 °C (0.05 mmHg)
Melting Pt: 130 °C
MDL Number: MFCD00005785
CAS Number: 136-95-8
EINECS: 205-268-4
Merck Index: 13,00423
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e0bb0cef1b9efa2b2f504d406a8b6d4f8426c72&vid=0e0bb0cef1b9efa2b2f504d406a8b6d4f8426c72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR