2-Aminobenzenesulfonic acid ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

A0266-25G A0266-100G
TCA0266-25GEA 49.61 CAD
TCA0266-25G TCA0266-100G
2-Aminobenzenesulfonic acid ≥99.0% (by HPLC, titration analysis)
2-Aminobenzenesulphonic acid

CAS Number: 88-21-1
MDL Number: MFCD00007705
Molecular Formula: C6H7NO3S
Molecular Weight: 173.19
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: o-Anilinesulfonic Acid, Orthanilic Acid, 2-Sulfoaniline

Formula: C₆H₇NO₃S
MDL Number: MFCD00007705
CAS Number: 88-21-1
EINECS: 201-810-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=571d23194b6b4dc5ccf529d56c9cc30ec1cdbbb4&vid=571d23194b6b4dc5ccf529d56c9cc30ec1cdbbb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR