3-Aminobenzene sulfonic acid ≥99.0%
Supplier: TCI America
Synonyms:
1-Aminobenzene-3-sulphonic acid, Metanilic acid, 3-Aminobenzenesulphonic acid, 3-Aminobenzenesulfonic acid, 1-Aminobenzene-3-sulfonic acid
A0265-500G
A0265-25G
TCA0265-500GEA
139.04
CAD
TCA0265-500G
TCA0265-25G
3-Aminobenzene sulfonic acid ≥99.0%
3-Aminobenzene sulphonic acid
CAS Number: 121-47-1
MDL Number: MFCD00065345
Molecular Formula: C6H7NO3S
Molecular Weight: 173.19
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Other Product Names / Synonyms: m-Anilinesulfonic Acid, Metanilic Acid, 3-Sulfoaniline
Formula:
C₆H₇NO₃S MW: 173.19 g/mol |
MDL Number:
MFCD00065345 CAS Number: 121-47-1 EINECS: 204-473-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...