Anthranilamide ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Aminobenzamide

A0262-25G A0262-500G A0262-100G
TCA0262-025GEA 153.26 CAD
TCA0262-025G TCA0262-500G TCA0262-100G
Anthranilamide ≥98.0%
Anthranilamide

CAS Number: 88-68-6
MDL Number: MFCD00007981
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 111


Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: Anthranilamide

Formula: C₇H₈N₂O
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 111 °C
Flash Pt: >185 °C
Storage Temperature: Ambient
MDL Number: MFCD00007981
CAS Number: 88-68-6
EINECS: 201-851-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f4e3fc9de163719a16392a398c12cb63cec5d73&vid=8f4e3fc9de163719a16392a398c12cb63cec5d73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR