4'-Aminoacetophenone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A0251-25G
A0251-100G
TCA0251-025GEA
102.7
CAD
TCA0251-025G
TCA0251-500G
TCA0251-100G
4'-Aminoacetophenone ≥98.0% (by titrimetric analysis)
4'-Aminoacetophenone
CAS Number: 99-92-3
MDL Number: MFCD00007896
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 293
Melting point (°C): 107
Formula:
C₈H₉NO Boiling Pt: 293 °C Melting Pt: 107 °C Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007896 CAS Number: 99-92-3 EINECS: 202-801-2 Merck Index: 13,00412 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...