Aluminon
Supplier: TCI America
Synonyms:
ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
CAS Number: 569-58-4
MDL Number: MFCD00040925
Molecular Weight: 473.44
Form: Crystal
Other Product Names / Synonyms: Ammonium Aurintricarboxylate, Aurintricarboxylic Acid Ammonium Salt
Formula:
C₂₂H₂₃N₃O₉ MW: 473.44 g/mol |
MDL Number:
MFCD00040925 CAS Number: 569-58-4 EINECS: 209-319-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...