Allyl phenyl ether ≥95.0%
Supplier: TCI America
A0234-25G
A0234-500G
A0234-100G
TCA0234-025GEA
94.8
CAD
TCA0234-025G
TCA0234-500G
TCA0234-100G
Allyl phenyl ether ≥95.0%
Allyl phenyl ether
CAS Number: 1746-13-0
MDL Number: MFCD00008644
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 62
Specific Gravity (20/20): 0.98
Formula:
H₂CCHCH₂OC₆H₅ Boiling Pt: 195 °C Density: 0.98 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00008644 CAS Number: 1746-13-0 EINECS: 217-125-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...