Diethyl allylmalonate ≥98.0%
Supplier: TCI America
A0231-500G
A0231-25G
TCA0231-500GEA
1560.41
CAD
TCA0231-500G
TCA0231-025G
Diethyl allylmalonate ≥98.0%
Diethyl allylmalonate
CAS Number: 2049-80-1
MDL Number: MFCD00009155
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 223
Flash Point (°C): 92
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: Allylmalonic Acid Diethyl Ester
Formula:
C₁₀H₁₆O₄ Boiling Pt: 223 °C Density: 1.01 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00009155 CAS Number: 2049-80-1 EINECS: 218-072-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...