β-Alanine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid

A0180-25G A0180-500G
TCA0180-025GEA 40.76 CAD
TCA0180-025G TCA0180-500G
β-Alanine ≥99.0% (by titrimetric analysis)
β-Alanine

CAS Number: 107-95-9
MDL Number: MFCD00008200
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 198


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 3-Aminopropionic Acid, H-beta-Ala-OH

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: 198 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0b97685041797d1c1f1ac667e5de3ab2607bf6e&vid=c0b97685041797d1c1f1ac667e5de3ab2607bf6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR