L-Alanine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: L-α-AlanineL(+)-AlanineAα-AlanineAlanineH-Ala-OH(S)-(+)-AlanineH-L-Ala-OH

A0179-250G A0179-25G
TCA0179-250GEA 170.04 CAD
TCA0179-250G TCA0179-025G
L-Alanine ≥99.0% (by titrimetric analysis)
L-Alanine

CAS Number: 56-41-7
MDL Number: MFCD00064410
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 315
Specific rotation [a]20/D: 14 deg (C=10, HCl)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: L-2-Aminopropionic Acid, H-Ala-OH

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 315 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00064410
CAS Number: 56-41-7
Merck Index: 13,00203
https://ca.vwr.com/stibo/web/std.lang.all/76/29/28927629.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR