DL-Alanine 98,5
Supplier: TCI America
Synonyms:
(±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
A0178-25G
A0178-500G
TCA0178-025GEA
26.86
CAD
TCA0178-025G
TCA0178-500G
DL-Alanine 98,5
DL-Alanine
CAS Number: 302-72-7
MDL Number: MFCD00064408
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Melting point (°C): 295
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-2-Aminopropionic Acid, H-DL-Ala-OH
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 189 Melting Pt: 79 Density: 1.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064408 CAS Number: 302-72-7 EINECS: 206-126-4 Merck Index: 13,00203 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...