Adonitol ≥97.0%
Supplier: TCI America
A0171-1G
A0171-25G
TCA0171-001GEA
36.34
CAD
TCA0171-001G
TCA0171-025G
Adonitol ≥97.0%
Adonitol
CAS Number: 488-81-3
MDL Number: MFCD00064291
Molecular Formula: C5H12O5
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 102
Product Categories: Bioscience, Sugar Alcohols, Ribose
Other Product Names / Synonyms: Adonitol
Formula:
HOH₂C(CH(OH))₃CH₂OH Melting Pt: 102 °C |
MDL Number:
MFCD00064291 CAS Number: 488-81-3 EINECS: 207-685-7 Merck Index: 13,00168 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...