Dimethyl adipate ≥99.0%
Supplier: TCI America
A0166-25ML
A0166-500ML
TCA0166-025MLEA
33.75
CAD
TCA0166-025ML
TCA0166-500ML
Dimethyl adipate ≥99.0%
Dimethyl adipate
CAS Number: 627-93-0
MDL Number: MFCD00008469
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 122
Freezing point (°C): 10
Specific Gravity (20/20): 1.06
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Adipic Acid Dimethyl Ester
Formula:
C₈H₁₄O₄ Boiling Pt: 110 °C Melting Pt: 10.3 °C Density: 1.06 (20 °C) Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008469 CAS Number: 627-93-0 EINECS: 211-020-6 Merck Index: 13,00163 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...