Bis(2-ethylhexyl)adipate ≥98.0%

Supplier: TCI America
Warning

A0163-500ML A0163-25ML
TCA0163-500MLEA 82.18 CAD
TCA0163-500ML TCA0163-025ML
Bis(2-ethylhexyl)adipate ≥98.0%
Bis(2-ethylhexyl) adipate

CAS Number: 103-23-1
MDL Number: MFCD00009496
Molecular Formula: C22H42O4
Molecular Weight: 370.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Melting point (°C): -70
Flash Point (°C): 205
Specific Gravity (20/20): 0.93


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Adipate (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Adipic Acid Dioctyl Ester, Adipic Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Adipate, Dioctyl Adipate, DOA

Formula: C₂₂H₄₂O₄
Boiling Pt: 214 °C
Melting Pt: –70 °C
Density: 0.93 (20 °C)
Flash Pt: 205 °C
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c23eff29cad10afea927708106924fb3669ce233&vid=c23eff29cad10afea927708106924fb3669ce233&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR