Diethyl adipate ≥99.0%

Supplier: TCI America

Synonyms: NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363

A0162-500ML A0162-25ML
TCA0162-500MLEA 88.31 CAD
TCA0162-500ML TCA0162-025ML
Diethyl adipate ≥99.0%
Diethyl adipate

CAS Number: 141-28-6
MDL Number: MFCD00009215
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 251
Melting point (°C): -20
Flash Point (°C): 127
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Adipic Acid Diethyl Ester

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 251 °C
Melting Pt: –20 °C
Density: 1.01 (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00009215
CAS Number: 141-28-6
EINECS: 205-477-0
Merck Index: 13,00163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66f17408cb1b5713ab3b6eacae6ab81f6172d2f4&vid=66f17408cb1b5713ab3b6eacae6ab81f6172d2f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR