Adipic acid 99,0

Supplier: TCI America
Warning

Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid

A0161-25G A0161-500G
TCA0161-025GEA 34.16 CAD
TCA0161-025G TCA0161-500G
Adipic acid 99,0
Adipic acid

CAS Number: 124-04-9
MDL Number: MFCD00004420
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 153
Flash Point (°C): 210


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Butanedicarboxylic Acid, Hexanedioic Acid

Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: 337
Melting Pt: 151
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a2aded8a4ef0c7e48f4f5b2cc3e709f19a8f311&vid=3a2aded8a4ef0c7e48f4f5b2cc3e709f19a8f311&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR