Adenosine-5'-monophosphoric acid (AMP) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, 5'-Adenylic acid, Adenosine 5'-monophosphate, Adenosine monophosphate, 5'-AMP

A0158-1G A0158-25G A0158-5G
TCA0158-001GEA 31.6 CAD
TCA0158-001G TCA0158-025G TCA0158-5G
Adenosine-5'-monophosphoric acid (AMP) ≥98.0% (by HPLC, titration analysis)
Adenosine-5'-monophosphoric acid (AMP)

CAS Number: 61-19-8
MDL Number: MFCD00005750
Molecular Formula: C10H14N5O7P
Molecular Weight: 347.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 200


Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: Adenosine 5'-Monophosphate, 5'-AMP

Formula: C₁₀H₁₆N₅O₈P
MW: 347.22 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00005750
CAS Number: 61-19-8
EINECS: 200-500-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e2c0f2dcc8f640b25fa3e4f1d390f6a2dc3b577&vid=8e2c0f2dcc8f640b25fa3e4f1d390f6a2dc3b577&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR