Adenine sulfate dihydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Aminopurine sulphate dihydrate, 7H-Purin-6-amine hemisulfate dihydrate, 6-Aminopurine sulfate dihydrate, 9H-Purin-6-amine hemisulfate dihydrate, Adenine hemisulphate dihydrate, 7H-Purin-6-amine hemisulphate dihydrate, 9H-Purin-6-amine hemisulphate dihydrate, Adenine hemisulfate dihydrate

A0151-25G A0151-100G
TCA0151-025GEA 154.84 CAD
TCA0151-025G TCA0151-100G
Adenine sulfate dihydrate ≥98.0% (by HPLC, titration analysis)
Adenine sulphate dihydrate

Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Cytokinins, Reagents for Inflammation & Immunity Disorder Models, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-Aminopurine Sulfate

Formula: C₁₀H₁₂N₁₀O₄S
MW: 404.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS Number: 321-30-2
EINECS: 206-286-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cefaddc8d33c228da17468285e75b961c7a3b69b&vid=cefaddc8d33c228da17468285e75b961c7a3b69b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR