Adenine hydrochloride hemihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Purin-6-amine hydrochloride hemihydrate, 1H-Purin-6-amine hydrochloride, 7H-Purin-6-amine hydrochloride hemihydrate, 6-Aminopurine Hydrochloride, Adenine Hydrochloride Hemihydrate, 6-Aminopurine hydrochloride hemihydrate, 7H-Purin-6-amine hydrochloride hydrate (2:2:1)
CAS Number: 2922-28-3
MDL Number: MFCD00075782
Molecular Formula: C5H5N5
Molecular Weight: 171.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-Aminopurine Hydrochloride
Formula:
C₅H₅N₅·HCl·½H₂O MW: 180.60 g/mol |
MDL Number:
MFCD11046306 CAS Number: 6055-72-7 EINECS: 220-867-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...