2-Ethylhexyl acrylate ≥99.0% stabilised
Supplier: TCI America
Synonyms:
Acrylic acid 2-ethylhexyl ester
Stabilization:
Stabilised with Stabilised
(stabilized with MEHQ)
CAS Number: 103-11-7
MDL Number: MFCD00009495
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Melting point (°C): -90
Flash Point (°C): 80
Specific Gravity (20/20): 0.89
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 2-Ethylhexyl Ester Monomer, Acrylic Acid Octyl Ester Monomer, Octyl Acrylate Monomer
Formula:
C₁₁H₂₀O₂ Boiling Pt: 215 °C Melting Pt: –90 °C Density: 0.89 (20 °C) Flash Pt: 80 °C |
MDL Number:
MFCD00009495 CAS Number: 103-11-7 EINECS: 203-080-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...