9-Acridone ≥98.0%

Supplier: TCI America

Synonyms: 9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone

A0133-25G A0133-5G
TCA0133-025GEA 380.78 CAD
TCA0133-025G TCA0133-5G
9-Acridone ≥98.0%
9-Acridone

CAS Number: 578-95-0
MDL Number: MFCD00005019
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 354

Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 354 °C
MDL Number: MFCD00005019
CAS Number: 578-95-0
EINECS: 209-434-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e5418d3b41c02d63ead8fb883be41ec96e4a59&vid=a0e5418d3b41c02d63ead8fb883be41ec96e4a59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR