trans-Aconitic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0127-25G A0127-500G
TCA0127-025GEA 102.86 CAD
TCA0127-025G TCA0127-500G
trans-Aconitic acid ≥98.0% (by HPLC, titration analysis)
trans-Aconitic acid

CAS Number: 4023-65-8
MDL Number: MFCD00002721
Molecular Formula: C6H6O6
Molecular Weight: 174.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180


Other Product Names / Synonyms: (E)-1-Propene-1,2,3-tricarboxylic Acid

Formula: C₆H₆O₆
Melting Pt: 180 °C
MDL Number: MFCD00002721
CAS Number: 4023-65-8
EINECS: 223-688-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be887b8b6ec29c6b910ab11c3a91b0b411b0ef5c&vid=be887b8b6ec29c6b910ab11c3a91b0b411b0ef5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR