N-Acetyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0120-1G
A0120-25G
TCA0120-001GEA
25.72
CAD
TCA0120-001G
TCA0120-025G
N-Acetyl-DL-tryptophan ≥98.0% (by HPLC, titration analysis)
N-Acetyl-DL-tryptophan
CAS Number: 87-32-1
MDL Number: MFCD00005644
Molecular Formula: C13H14N2O3
Molecular Weight: 246.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 205
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-DL-Trp-OH
Formula:
C₁₃H₁₄N₂O₃ Melting Pt: 205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005644 CAS Number: 87-32-1 EINECS: 201-739-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...