4-Acetylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0113-25G A0113-250G
TCA0113-025GEA 75.84 CAD
TCA0113-025G TCA0113-250G
4-Acetylpyridine ≥98.0% (by GC, titration analysis)
4-Acetylpyridine

CAS Number: 1122-54-9
MDL Number: MFCD00006433
Molecular Formula: C7H7NO
Molecular Weight: 121.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 214
Melting point (°C): 16
Flash Point (°C): 104
Specific Gravity (20/20): 1.10

Formula: C₇H₇NO
Boiling Pt: 214 °C
Melting Pt: 16 °C
Density: 1.10 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006433
CAS Number: 1122-54-9
EINECS: 214-350-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15b357eb4016faac725e17f27cff5daec9281a4f&vid=15b357eb4016faac725e17f27cff5daec9281a4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR