2-Acetylpyridine ≥99.0%

Supplier: TCI America

Synonyms: 1-(Pyridin-2-yl)ethanone, Methyl 2-pyridyl ketone

A0111-250ML A0111-25ML
TCA0111-250MLEA 287.56 CAD
TCA0111-250ML TCA0111-025ML
2-Acetylpyridine ≥99.0%
2-Acetylpyridine

CAS Number: 1122-62-9
MDL Number: MFCD00006303
Molecular Formula: C7H7NO
Molecular Weight: 121.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 73
Specific Gravity (20/20): 1.08

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 189 °C
Melting Pt: ∼10 °C
Density: 1.08 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006303
CAS Number: 1122-62-9
EINECS: 214-355-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d782f2c7611b1821dab78b2549670fa86123e8c&vid=2d782f2c7611b1821dab78b2549670fa86123e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR