4'-Aminoacetanilide ≥98.0%

Supplier: TCI America
Warning

Synonyms: N-(4-Aminophenyl)acetamide

A0106-500G A0106-25G
TCA0106-500GEA 320.74 CAD
TCA0106-500G TCA0106-025G
4'-Aminoacetanilide ≥98.0%
4'-Aminoacetanilide

CAS Number: 122-80-5
MDL Number: MFCD00007853
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 166
Flash Point (°C): 150


Other Product Names / Synonyms: 4-Acetamidoaniline, N-Acetyl-1,4-phenylenediamine

Formula: C₈H₁₀N₂O
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 166 °C
Density: 1.27 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007853
CAS Number: 122-80-5
EINECS: 204-576-6
Merck Index: 13,00410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa4292b00f5faa174260da01e41df4acaaf651d&vid=daa4292b00f5faa174260da01e41df4acaaf651d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR