3'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0104-500G A0104-25G
TCA0104-500GEA 429.76 CAD
TCA0104-500G TCA0104-025G
3'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
3'-Hydroxyacetophenone

CAS Number: 121-71-1
MDL Number: MFCD00002298
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 153
Melting point (°C): 94


Other Product Names / Synonyms: 3-Acetophenol, 3-Acetylphenol, 3-Hydroxyphenyl Methyl Ketone

Formula: C₈H₈O₂
Boiling Pt: 153 °C
Melting Pt: 94 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1
EINECS: 204-494-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f7646a79a1b4fa6aafad71ae8e25d77308ee570&vid=3f7646a79a1b4fa6aafad71ae8e25d77308ee570&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR