N-Acetylglycine(Ac-Gly-OH) ≥99.0%

Supplier: TCI America

Synonyms: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid

A0093-25G A0093-500G A0093-100G
TCA0093-025GEA 33.18 CAD
TCA0093-025G TCA0093-500G TCA0093-100G
N-Acetylglycine(Ac-Gly-OH) ≥99.0%
N-Acetylglycine (Ac-Gly-OH)

CAS Number: 543-24-8
MDL Number: MFCD00004275
Molecular Formula: C4H7NO3
Molecular Weight: 117.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 207


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Acetamidoacetic Acid, Aceturic Acid, Ac-Gly-OH

Formula: C₄H₇NO₃
MW: 117.1 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00004275
CAS Number: 543-24-8
EINECS: 208-839-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed83a21dc727fc21a98ef0a3d592e39f8dbbfb5&vid=9ed83a21dc727fc21a98ef0a3d592e39f8dbbfb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR