N-Acetyl-β-D-glucosamine ≥98.0%

Supplier: TCI America

Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

A0092-25G A0092-500G A0092-100G
TCA0092-025GEA 82.29 CAD
TCA0092-025G TCA0092-500G TCA0092-100G
N-Acetyl-β-D-glucosamine ≥98.0%
N-Acetyl-β-D-glucosamine

CAS Number: 7512-17-6
MDL Number: MFCD00136044
Molecular Formula: C8H15NO6
Molecular Weight: 221.21
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 205
Specific rotation [a]20/D: 40.5 deg (C=1, H2O)


Product Categories: Bioscience, Glucosamine, Aminosugars
Other Product Names / Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose

Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00136044
CAS Number: 7512-17-6
EINECS: 231-368-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c84d4d1267d62db60de1a9764795c460ae962a47&vid=c84d4d1267d62db60de1a9764795c460ae962a47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR