2-Amino-5-chlorobenzophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone
A0079-25G
A0079-500G
TCA0079-025GEA
74.26
CAD
TCA0079-025G
TCA0079-500G
2-Amino-5-chlorobenzophenone ≥98.0%
2-Amino-5-chlorobenzophenone
CAS Number: 719-59-5
MDL Number: MFCD00007839
Molecular Formula: C13H10ClNO
Molecular Weight: 231.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 98
Formula:
C₁₃H₁₀ClNO Boiling Pt: 424 °C (1013 hPa) Melting Pt: 98 °C |
MDL Number:
MFCD00007839 CAS Number: 719-59-5 EINECS: 211-949-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...