3-Acetoxyindole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A0068-1G A0068-5G
TCA0068-001GEA 140.62 CAD
TCA0068-001G TCA0068-005G
3-Acetoxyindole ≥98.0% (by HPLC, total nitrogen)
3-Acetoxyindole

CAS Number: 608-08-2
MDL Number: MFCD00014561
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 3-Acetoxyindole, 3-Indolyl Acetate

Formula: C₁₀H₉NO₂
Melting Pt: 130 °C
MDL Number: MFCD00014561
CAS Number: 608-08-2
EINECS: 210-154-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1093a8f1d8a57262963d72c30e066a65eaab0bbb&vid=1093a8f1d8a57262963d72c30e066a65eaab0bbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR