4'-Methylacetanilide ≥98.0%

Supplier: TCI America

Synonyms: p-Acetotoluidid, p-Acetotoluidide, N-(4-Methylphenyl)acetamide

A0064-500G A0064-25G A0064-100G
TCA0064-500GEA 1090.2 CAD
TCA0064-500G TCA0064-025G TCA0064-100G
4'-Methylacetanilide ≥98.0%
4'-Methylacetanilide

CAS Number: 103-89-9
MDL Number: MFCD00008677
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 307
Melting point (°C): 150
Flash Point (°C): 169


Other Product Names / Synonyms: N-Acetyl-p-toluidine, 4'-Methylacetanilide

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 307 °C
Melting Pt: 150 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00008677
CAS Number: 103-89-9
EINECS: 203-155-4
Merck Index: 13,00076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37bb0bc01bb89314fc28572d2bb255e578069ce8&vid=37bb0bc01bb89314fc28572d2bb255e578069ce8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR