Dimethyl-3-oxoglutarate ≥95.0%
Supplier: TCI America
Synonyms:
Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate
CAS Number: 1830-54-2
MDL Number: MFCD00008462
Molecular Formula: C7H10O5
Molecular Weight: 174.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: 1,3-Acetonedicarboxylic Acid Dimethyl Ester, Dimethyl 3-Oxoglutarate, 3-Oxoglutaric Acid Dimethyl Ester, Dimethyl 3-Oxopentanedioate, 3-Oxopentanedioic Acid Dimethyl Ester
Formula:
C₇H₁₀O₅ MW: 174.15 g/mol Boiling Pt: 150 °C Density: 1.21 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008462 CAS Number: 1830-54-2 EINECS: 217-385-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...