Diethyl-3-oxoglutarate ≥95.0%
Supplier: TCI America
A0055-25ML
A0055-500ML
TCA0055-025MLEA
79
CAD
TCA0055-025ML
TCA0055-500ML
Diethyl-3-oxoglutarate ≥95.0%
Diethyl 3-oxoglutarate
CAS Number: 105-50-0
MDL Number: MFCD00009200
Molecular Formula: C9H14O5
Molecular Weight: 202.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 250
Flash Point (°C): 71
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: 1,3-Acetonedicarboxylic Acid Diethyl Ester, Diethyl 3-Oxoglutarate, 3-Oxoglutaric Acid Diethyl Ester, Diethyl 3-Oxopentanedioate, 3-Oxopentanedioic Acid Diethyl Ester
Formula:
C₉H₁₄O₅ Boiling Pt: 250 °C Density: 1.11 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00009200 CAS Number: 105-50-0 EINECS: 203-302-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...