1-Acetylnaphthalene ≥98.0%

Supplier: TCI America

Synonyms: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

A0052-25G A0052-500G
TCA0052-025GEA 52.14 CAD
TCA0052-025G TCA0052-500G
1-Acetylnaphthalene ≥98.0%
1-Acetylnaphthalene

CAS Number: 941-98-0
MDL Number: MFCD00004013
Molecular Formula: C12H10O
Molecular Weight: 170.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 298
Flash Point (°C): 110
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 1-Acetylnaphthalene, Methyl 1-Naphthyl Ketone

Formula: C₁₂H₁₀O
Boiling Pt: 298 °C
Melting Pt: 9…11 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004013
CAS Number: 941-98-0
EINECS: 213-384-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69317198d2e8c67b2c4e4ecd7023a0a517cf2182&vid=69317198d2e8c67b2c4e4ecd7023a0a517cf2182&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR