Acetoacetanilide ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America

A0046-25G A0046-500G
TCA0046-025GEA 31.6 CAD
TCA0046-025G TCA0046-500G
Acetoacetanilide ≥99.0% (by HPLC, total nitrogen)
Acetoacetanilide

CAS Number: 102-01-2
MDL Number: MFCD00008780
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method: >99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 85
Flash Point (°C): 185

Formula: C₁₀H₁₁NO₂
Boiling Pt: 129 °C (32 mmHg)
Melting Pt: 85 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00008780
CAS Number: 102-01-2
EINECS: 202-996-4
Merck Index: 13,00059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f91d6aada24729f8c2909e6344b0f7f0b6ceecf8&vid=f91d6aada24729f8c2909e6344b0f7f0b6ceecf8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR