Octyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic acid octyl ester

A0042-25ML A0042-500ML
TCA0042-025MLEA 35.28 CAD
TCA0042-025ML TCA0042-500ML
Octyl acetate ≥98.0%
Octyl acetate

CAS Number: 112-14-1
MDL Number: MFCD00009562
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 95
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid n-Octyl Ester

Formula: C₁₀H₂₀O₂
Boiling Pt: 210 °C
Melting Pt: –38.5 °C
Density: 0.87 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00009562
CAS Number: 112-14-1
EINECS: 203-939-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8fb9d5807b1d67cb3c1e42b89204f66134d09de&vid=5f4e5317e303701d44f625e6818c40a68d1fb98b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR