Hexyl acetate ≥99.0%

Supplier: TCI America

Synonyms: Acetic acid hexyl ester

A0032-500ML A0032-25ML
TCA0032-500MLEA 88.48 CAD
TCA0032-500ML TCA0032-25ML
Hexyl acetate ≥99.0%
Hexyl acetate

CAS Number: 142-92-7
MDL Number: MFCD00009524
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 169
Flash Point (°C): 62
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid Hexyl Ester

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 169 °C
Melting Pt: –80 °C
Density: 0.87 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009524
CAS Number: 142-92-7
EINECS: 205-572-7
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e61aa227bb687c7418ea33167ed1e09fd3b7325&vid=c3d7e3b7722c4f2e6cba91efb93e3a1d74c29e86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR